Learn More
Brady™ Conical Shaped Drain Plugs
Catalog No. p-4955345 Shop All Brady Corporation Products
Click to view available options
Diameter (Metric):
Diameter (English):
Prevents spills and leaks from entering the drainage system
  • Conocal shaped made of chemical-resistant PVC
  • Features a “metal eye” for ease of insertion and removal
  • Available in four sizes to fit 2, 3, 4 and 5 in. drains
  • Rinsed and re-used
Specifications
Color
Yellow
Material
PVC
Type
6 in. Drain Plug
Shape
Conical
Diameter (English)
6 in.
Diameter (Metric)
15.2 cm