Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4H-Cyclopenta[2,1-b:3,4-b′]dithiophene 97.0+%, TCI America™
SDP

Catalog No. p-7125787 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 389-58-2
Molecular Formula C9H6S2
Molecular Weight (g/mol) 178.27
MDL Number MFCD20133900
InChI Key UITASDKJJNYORO-UHFFFAOYSA-N
Synonym 4H-Cyclopenta[1,2-b:5,4-b′C]dithiophene
PubChem CID 136232
IUPAC Name 3,11-dithiatricyclo[6.3.0.0²,⁶]undeca-1(8),2(6),4,9-tetraene
SMILES C1C2=C(SC=C2)C2=C1C=CS2

Specifications

Melting Point 73°C
Quantity 200 mg
Formula Weight 178.27
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4H-Cyclopenta[2,1-b:3,4-b′]dithiophene
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.