Learn More
Accuflow Systems Sterile Empty Vial (SEV), 5.0 mL, 20 mm clear glass with silver cap - 25/pack
ALK Sterile Empty Vials (SEV) feature Type 1 glass USP borosilicate, and a latex free stopper. ALK SEVs are produced at a cGMP FDA Manufacturer and USP tested for sterility and endotoxins. Certificate of Analysis available for all production lots and vials are aseptically assembled in an ISO 5 (class 100) cleanroom.