Learn More
Sklar Instruments Scissors, Merit Mayo Dissecting Scissors, Sklar 5-1/2 in., Blunt/Blunt, Curved, Stainless Steel, EA.
SDP
Supplier:  Sklar Instruments 97312
Encompass_Preferred
Scissors, Merit Mayo Dissecting Scissors, Sklar, 5-1/2 in., Blunt/Blunt, Curved, Stainless Steel, EA. Merit Mayo Dissecting Scissors are a versatile instrument that can be used in multiple procedures and multiple settings. They are available straight or curved in a variety of lengths with a standard beveled blade. While the curved instrument is most commonly utilized for cutting or dissecting dense or deep tissue, and the straight version is typically used for cutting surface tissue or sutures, both may be used interchangeably depending on procedure and physician preference. This product is straight with smooth, blunt/blunt tips and a length of 5-1/2 inches.
Catalog No. 50-109-4113
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.