Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,10,15,20-Tetrakis(2,6-dichlorophenyl)porphyrin 95.0+%, TCI America™
SDP

Catalog No. T1438100MG Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:

Chemical Identifiers

CAS

37083-37-7

Molecular Formula

C44H22Cl8N4

Molecular Weight (g/mol)

890.288

MDL Number

MFCD00191500

InChI Key

LOKNIYQQLQPXMC-UHFFFAOYSA-N

Synonym

5,10,15,20-Tetrakis(2,6-dichlorophenyl)porphine

PubChem CID

481556

IUPAC Name

5,10,15,20-tetrakis(2,6-dichlorophenyl)-21,22-dihydroporphyrin

SMILES

C1=CC(=C(C(=C1)Cl)C2=C3C=CC(=C(C4=CC=C(N4)C(=C5C=CC(=N5)C(=C6C=CC2=N6)C7=C(C=CC=C7Cl)Cl)C8=C(C=CC=C8Cl)Cl)C9=C(C=CC=C9Cl)Cl)N3)Cl

Specifications

Color

Purple

Quantity

100 mg

Formula Weight

890.29

Percent Purity

≥95.0% (HPLC)

Physical Form

Crystalline Powder

Chemical Name or Material

5,10,15,20-Tetrakis(2,6-dichlorophenyl)porphyrin

TSCA

No

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.