Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,15-Diphenylporphyrin 90.0+%, TCI America™
SDP

Catalog No. D2860100MG Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:

Chemical Identifiers

CAS

22112-89-6

Molecular Formula

C32H22N4

Molecular Weight (g/mol)

462.556

InChI Key

QIBKIAFNCVIIMG-UHFFFAOYSA-N

Synonym

5,15-Diphenyl-21H,23H-porphine

PubChem CID

10895852

IUPAC Name

10,20-diphenyl-21,22-dihydroporphyrin

SMILES

C1=CC=C(C=C1)C2=C3C=CC(=CC4=CC=C(N4)C(=C5C=CC(=N5)C=C6C=CC2=N6)C7=CC=CC=C7)N3

Specifications

Color

Blue-Purple

Quantity

100 mg

Formula Weight

462.56

Percent Purity

≥90.0% (HPLC)

Physical Form

Crystalline Powder

Chemical Name or Material

5,15-Diphenylporphyrin

TSCA

No

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.