Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(2-Bromophenyl)-3-methyl-1,2,4-oxadiazole 97+%, Thermo Scientific™

Catalog No. p-4510707
Click to view available options
Quantity:
250 mg
1 g
5 g

Chemical Identifiers

CAS 375857-64-0
Molecular Formula C9H7BrN2O
Molecular Weight (g/mol) 239.072
InChI Key NYAFKUCVAVUCHW-UHFFFAOYSA-N
PubChem CID 18001310
IUPAC Name 5-(2-bromophenyl)-3-methyl-1,2,4-oxadiazole
SMILES CC1=NOC(=N1)C2=CC=CC=C2Br

Specifications

Assay Percent Range 97%
Quantity 250 mg
Synonym 5-2-bromophenyl-3-methyl-1,2,4-oxadiazole, 1,2,4-oxadiazole, 5-2-bromophenyl-3-methyl, 5-2-bromophenyl-3-methyl-1,2,4 oxadiazole, 1,2,4-oxadiazole,5-2-bromophenyl-3-methyl
Formula Weight 239.07
Percent Purity 97%
Chemical Name or Material 5-(2-Bromophenyl)-3-methyl-1, 2, 4-oxadiazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.