Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
5,5-Dibromomeldrum′s Acid (=5,5-Dibromo-2,2-dimethyl-4,6-dioxy-1,3-dioxane) 98.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
5 g
25 g
Specifications
Specifications
Melting Point | 75°C |
Color | White-Yellow |
Quantity | 5 g |
Formula Weight | 301.92 |
Percent Purity | ≥98.0% (T) |
Physical Form | Crystalline Powder |
Chemical Name or Material | 5,5-Dibromomeldrum′s Acid (=5,5-Dibromo-2,2-dimethyl-4,6-dioxy-1,3-dioxane) |
Safety and Handling
TSCA | No |
Recommended Storage | Freezer |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction