Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,7-Dichlorokynurenic acid, Tocris Bioscience™

Catalog No. p-4783191 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent NMDA antagonist; acts at glycine site

Potent antagonist at the glycine site of the NMDA receptor (Ki = 79 nM vs. [3H]-glycine).

Sodium Salt also available.

Chemical Identifiers

CAS 131123-76-7
Molecular Formula C10H5Cl2NO3
Molecular Weight (g/mol) 258.054
InChI Key BGKFPRIGXAVYNX-UHFFFAOYSA-N
Synonym 5,7-dichlorokynurenic acid, 5,7-dichloro-4-hydroxyquinoline-2-carboxylic acid, 5,7-dcka, 5,7-dichloroquinurenic acid, dcka, 5,7-dichorokynurenic acid, unii-t61ork73py, 5,7-dichloro-4-hydroxy-2-quinolinecarboxylic acid, t61ork73py, chembl50267
PubChem CID 1779
IUPAC Name 5,7-dichloro-4-oxo-1H-quinoline-2-carboxylic acid
SMILES C1=C(C=C2C(=C1Cl)C(=O)C=C(N2)C(=O)O)Cl

Specifications

Quantity 10 mg
Formula Weight 258.06
Percent Purity >98%
Chemical Name or Material 5, 7-Dichlorokynurenic acid
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.