Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-1-methyl-1H-pyrazole-4-carboxamide, Thermo Scientific™

Catalog No. KM00775CB
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 18213-75-7
Molecular Formula C5H8N4O
Molecular Weight (g/mol) 140.146
MDL Number MFCD00111808
InChI Key JGSQVTVXGXOSCH-UHFFFAOYSA-N
PubChem CID 265696
IUPAC Name 5-amino-1-methylpyrazole-4-carboxamide
SMILES CN1C(=C(C=N1)C(=O)N)N

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 5-amino-1-methyl-1h-pyrazole-4-carboxamide, 1h-pyrazole-4-carboxamide, 5-amino-1-methyl, pubchem22392, 5-amino-1-methyl-4-pyrazolecarboxamide, 5-azanyl-1-methyl-pyrazole-4-carboxamide, 1-methyl-5-amino-1h-pyrazole-4-carboxamide, 1h-pyrazole-4-carboxamide,5-amino-1-methyl, 5-imino-1-methyl-2h-pyrazole-4-carboxamide, 3-imino-2-methyl-2,3-dihydro-1h-pyrazole-4-carboxamide
Formula Weight 140.15
Percent Purity 95%
Chemical Name or Material 5-Amino-1-methyl-1H-pyrazole-4-carboxamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.