Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-1H-[1,2,4]-triazole-3-carboxylic acid methyl ester, 96%, Thermo Scientific™

Catalog No. AC437690010
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 3641-14-3
Molecular Formula C4H6N4O2
Molecular Weight (g/mol) 142.12
InChI Key OSZNPKRMPBUQLB-UHFFFAOYSA-N
Synonym methyl 5-amino-1h-1,2,4-triazole-3-carboxylate, methyl 5-amino-4h-1,2,4-triazole-3-carboxylate, methyl 5-amino-4h-1,2,4 triazole-3-carboxylate, 1h-1,2,4-triazole-3-carboxylic acid, 5-amino-, methyl ester, 5-amino-1h-1,2,4-triazole-3-carboxylic acid methyl ester, methyl 5-amino-1,2,4-triazole-3-carboxylate, methyl-5-amino-1h-1,2,4-triazole-3-carboxylate, 5-amino-1h-1,2,4 triazole-3-carboxylic acid methyl ester, methyl 5-amino-2h-1,2,4-triazole-3-carboxylate
PubChem CID 77201
IUPAC Name methyl 3-amino-1H-1,2,4-triazole-5-carboxylate
SMILES COC(=O)C1=NC(=NN1)N

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 212°C to 213°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 142.12
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-1H-[1,2,4]-triazole-3-carboxylic acid methyl ester
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 222-869-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.