Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-2-methylbenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. p-7117868 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 2840-04-2
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD06208351
InChI Key FSXVZWAWYKMFMX-UHFFFAOYSA-N
Synonym 5-amino-2-methyl-benzoic acid, 5-amino-2-methyl benzoic acid, 2-methyl-5-aminobenzoic acid, 5-amino-o-toluic acid, 5-amino-2-methylbenzoicacid, benzoic acid, 5-amino-2-methyl, pubchem4980, 3-amino-6-methylbenzoic acid, ksc494q3t, 2-methyl-5-amino benzoic acid
PubChem CID 10374614
IUPAC Name 5-amino-2-methylbenzoic acid
SMILES CC1=C(C=C(C=C1)N)C(=O)O

Specifications

Melting Point 197°C
Quantity 5 g
Formula Weight 151.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-2-methylbenzoic Acid
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.