Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-3-methyl-1-phenylpyrazole, 97%, Thermo Scientific Chemicals

Catalog No. p-4813185
Click to view available options
Quantity:
5 g
25 g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1131-18-6
Molecular Formula C10H11N3
Molecular Weight (g/mol) 173.21
InChI Key FMKMKBLHMONXJM-UHFFFAOYSA-N
Synonym 3-methyl-1-phenyl-1h-pyrazol-5-amine, 5-amino-3-methyl-1-phenylpyrazole, 1h-pyrazol-5-amine, 3-methyl-1-phenyl, 1-phenyl-3-methyl-5-aminopyrazole, 3-methyl-1-phenylpyrazol-5-ylamine, 5-methyl-2-phenyl-2h-pyrazol-3-ylamine, 1-phenyl-3-methyl-5-amino pyrazole, unii-x6087geb2p, pyrazole, 5-amino-3-methyl-1-phenyl, 1h-pyrazole-5-amine, 3-methyl-1-phenyl
PubChem CID 70801
IUPAC Name 5-methyl-2-phenylpyrazol-3-amine
SMILES CC1=NN(C(=C1)N)C2=CC=CC=C2

Specifications

Melting Point 113°C to 115°C
Boiling Point 333°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 173.21
Percent Purity 97%
Chemical Name or Material 5-Amino-3-methyl-1-phenylpyrazole
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e
EINECSNumber 214-463-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.