Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-4H-1,2,4-triazole-3-thiol, tech., Thermo Scientific™

Catalog No. SB00513DA
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 16691-43-3
Molecular Formula C2H4N4S
Molecular Weight (g/mol) 116.14
MDL Number MFCD00005231
InChI Key WZUUZPAYWFIBDF-UHFFFAOYSA-N
Synonym 3-amino-5-mercapto-1,2,4-triazole, iminothiourazole, 3-imino-5-thiourazole, 3-amino-1h-1,2,4-triazole-5-thiol, 5-amino-4h-1,2,4-triazole-3-thiol, 3-amino-1,2,4-triazole-5-thiol, s-triazole-3-thiol, 5-amino, 3h-1,2,4-triazole-3-thione, 5-amino-1,2-dihydro, 5-amino-1,2-dihydro-3h-1,2,4-triazole-3-thione, unii-353f2d8l1z
PubChem CID 2723869
IUPAC Name 5-amino-1,2-dihydro-1,2,4-triazole-3-thione
SMILES NC1=NC(=S)NN1

Specifications

Quantity 1 g
Formula Weight 116.15
Chemical Name or Material 5-Amino-4H-1, 2, 4-triazole-3-thiol
Hazard Category Warning
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.