Learn More
AdipoGen 5-Aminolevulinic acid HCl
SDP
Supplier:  AdipoGen CDXA0364M500
Encompass_Preferred
Chemical. CAS 5451-09-2. Formula C5H9NO3 . HCl. MW 131.13 . 36.46. 5-Aminolevulinic acid 5-ALA is a non-proteinogenic five carbon amino acid, which is the precursor for the biosynthesis of tetrapyrrole compounds, such as heme, chlorophyll and vitamin B12 and has broad applications in the medical and agricultural fields. The conversion of 5-ALA to protoporphyrins within tissues produces a photosensitive target that produces reactive oxygen species upon exposure to light. In this way, it is used in photodynamic therapy for a range of dermatological conditions, cancers, and other diseases. Oral administration of 5-ALA leads to the preferential accumulation of the fluorescent molecule protoporphyrin IX within certain types of cancer cells. This allows fluorescence-based identification of tumor tissue for accurate resection of diseased tissue.
Catalog No. 50-256-1741
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.