Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Benzyloxy-2-bromo-4-methoxybenzoic Acid 97.0+%, TCI America™
SDP

Catalog No. p-7122879 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 24958-42-7
Molecular Formula C15H13BrO4
Molecular Weight (g/mol) 337.17
MDL Number MFCD03695499
InChI Key FBTDEMXQQIWHKX-UHFFFAOYSA-N
Synonym 5-Benzyloxy-2-bromo-p-anisic Acid
PubChem CID 11056839
IUPAC Name 5-(benzyloxy)-2-bromo-4-methoxybenzoic acid
SMILES COC1=C(OCC2=CC=CC=C2)C=C(C(O)=O)C(Br)=C1

Specifications

Melting Point 195°C
Quantity 1 g
Formula Weight 337.17
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Benzyloxy-2-bromo-4-methoxybenzoic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.