Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2,3-difluorophenol 98.0+%, TCI America™
SDP

Catalog No. p-7123574 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 186590-26-1
Molecular Formula C6H3BrF2O
Molecular Weight (g/mol) 208.99
MDL Number MFCD01631350
InChI Key QAHCQGXGAYRHHW-UHFFFAOYSA-N
Synonym 2,3-difluoro-5-bromophenol, phenol, 5-bromo-2,3-difluoro, 5-bromo-2,3-difluoro-phenol, 3-bromo-5,6-difluorophenol, pubchem2843, acmc-209eoh, ksc494o6r, 5-bromo-2,3-difluoro phenol, phenol,5-bromo-2,3-difluoro, 2.3-difluoro-5-bromophenol
PubChem CID 2773307
IUPAC Name 5-bromo-2,3-difluorophenol
SMILES C1=C(C=C(C(=C1F)F)O)Br

Specifications

Color Yellow
Quantity 5 g
Formula Weight 208.99
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Bromo-2,3-difluorophenol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.