Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-fluorophenol 98.0+%, TCI America™
SDP

Catalog No. p-7119925 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 112204-58-7
Molecular Formula C6H4BrFO
Molecular Weight (g/mol) 190.999
MDL Number MFCD07782066
InChI Key YPTHSYKJDRMAJY-UHFFFAOYSA-N
Synonym 2-fluoro-5-bromophenol, phenol, 5-bromo-2-fluoro, 2-fluoro-5-bromo phenol, 5-bromo-2-fluoro-phenol, 3-bromo-6-fluorophenol, 3-bromo-6-fluoro-phenol, acmc-2099dd, ksc493i2d
PubChem CID 183421
IUPAC Name 5-bromo-2-fluorophenol
SMILES C1=CC(=C(C=C1Br)O)F

Specifications

Color Yellow
Boiling Point 54°C
Quantity 5 g
Formula Weight 191.00
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 5-Bromo-2-fluorophenol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.