Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-hydroxy-3-nitropyridine 98.0+%, TCI America™
SDP

Catalog No. p-7119728 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 15862-34-7
Molecular Formula C5H3BrN2O3
Molecular Weight (g/mol) 218.994
InChI Key WXRLCVUDLFFTFF-UHFFFAOYSA-N
Synonym 5-bromo-2-hydroxy-3-nitropyridine, 5-bromo-3-nitropyridin-2-ol, 5-bromo-3-nitro-2-pyridinol, 5-bromo-3-nitro-2-pyridone, 5-bromo-3-nitropyridin-2 1h-one, 5-bromo-3-nitro-pyridin-2-ol, 3-bromo-6-hydroxy-5-nitropyridine, 5-bromo-3-nitro-2 1h-pyridinone, 5-bromo-3-nitro-2-hydroxypyridine, 2-hydroxy-3-nitro-5-bromopyridine
PubChem CID 85147
IUPAC Name 5-bromo-3-nitro-1H-pyridin-2-one
SMILES C1=C(C(=O)NC=C1Br)[N+](=O)[O-]

Specifications

Melting Point 247°C
Color Yellow
Quantity 1 g
Formula Weight 218.99
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-hydroxy-3-nitropyridine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.