Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-methoxyphenylboronic Acid (contains varying amounts of Anhydride) 98.0+%, TCI America™
SDP

Catalog No. p-7123999 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 89694-45-1
Molecular Formula C7H8BBrO3
Molecular Weight (g/mol) 230.852
MDL Number MFCD00093409
InChI Key FVRLSHRAYPFXRQ-UHFFFAOYSA-N
Synonym 5-bromo-2-methoxyphenyl boronic acid, 5-bromo-2-methoxybenzeneboronic acid, 4-bromoanisole-2-boronic acid, 3-bromo-6-methoxyphenylboronic acid, boronic acid,b-5-bromo-2-methoxyphenyl, 5-bromo-2-methoxyphenylboronicacid, rarechem ah pb 0045, acmc-209r2g, amtb789, 2-methoxy-5-bromophenylboronic acid
PubChem CID 4532730
IUPAC Name (5-bromo-2-methoxyphenyl)boronic acid
SMILES B(C1=C(C=CC(=C1)Br)OC)(O)O

Specifications

Melting Point 131°C
Color White-Yellow
Quantity 1 g
Formula Weight 230.85
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-methoxyphenylboronic Acid (contains varying amounts of Anhydride)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.