Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-methylbenzonitrile 98.0+%, TCI America™
SDP

Catalog No. p-7123764 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 156001-51-3
Molecular Formula C8H6BrN
Molecular Weight (g/mol) 196.047
MDL Number MFCD08061920
InChI Key WNVUTFDOGUGEIS-UHFFFAOYSA-N
Synonym 4-Bromo-2-cyanotoluene
PubChem CID 12994004
IUPAC Name 5-bromo-2-methylbenzonitrile
SMILES CC1=C(C=C(C=C1)Br)C#N

Specifications

Melting Point 50°C
Color White-Yellow
Quantity 1 g
UN Number 3439
Formula Weight 196.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-methylbenzonitrile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.