Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-thiophenesulfonamide 97.0+%, TCI America™
SDP

Catalog No. p-7119761 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 53595-65-6
Molecular Formula C4H4BrNO2S2
Molecular Weight (g/mol) 242.11
MDL Number MFCD00067990
InChI Key WXJQQLDICAOBJB-UHFFFAOYSA-N
Synonym 5-bromo-2-thiophenesulfonamide, 5-bromo-thiophene-2-sulfonic acid amide, 2-thiophenesulfonamide, 5-bromo, 5-bromothiophene-2-sulfonic acid amide, 5-bromothiophene-2-sulphonamide, 2-bromo thiophene-5-sulfonamide, 5-bromothiophene-2-sulfonylamine, pubchem7369, maybridge1_004885, acmc-1b1sa
PubChem CID 1241304
IUPAC Name 5-bromothiophene-2-sulfonamide
SMILES NS(=O)(=O)C1=CC=C(Br)S1

Specifications

Melting Point 141°C
Color Yellow
Quantity 5 g
Formula Weight 242.11
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-thiophenesulfonamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.