Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromoindole-3-carboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. p-7123751 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 10406-06-1
Molecular Formula C9H6BrNO2
Molecular Weight (g/mol) 240.056
MDL Number MFCD05664007
InChI Key JVZMBSGNSAHFCY-UHFFFAOYSA-N
Synonym 5-bromoindole-3-carboxylic acid, 5-bromo-indole-3-carboxylic acid, 5-bromo-3-indolecarboxylic acid, 1h-indole-3-carboxylic acid,5-bromo, 1h-indole-3-carboxylic acid, 5-bromo, 5-bromo-1h-indole-3-carboxylicacid, pubchem7982, acmc-1btmm, ksc173q4r, 5-bromo-3-indolecarboxylicacid
PubChem CID 7018243
IUPAC Name 5-bromo-1H-indole-3-carboxylic acid
SMILES C1=CC2=C(C=C1Br)C(=CN2)C(=O)O

Specifications

Color White-Yellow
Boiling Point 240°C
Quantity 1 g
Formula Weight 240.06
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromoindole-3-carboxylic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.