Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(Bromomethyl)-2,1,3-benzothiadiazole, 95+%, Thermo Scientific™

Catalog No. CC09208DA
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 65858-50-6
Molecular Formula C7H5BrN2S
Molecular Weight (g/mol) 229.095
InChI Key JEPACAAYCVWCFI-UHFFFAOYSA-N
PubChem CID 591004
IUPAC Name 5-(bromomethyl)-2,1,3-benzothiadiazole
SMILES C1=CC2=NSN=C2C=C1CBr

Specifications

Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 1 g
Synonym 5-bromomethyl-2,1,3-benzothiadiazole, 5-bromomethyl benzo c 1,2,5 thiadiazole, 5-bromomethylbenzo-2,1,3-thiadiazole, 5-bromomethyl-benzo 1,2,5 thiadiazole, 2,1,3-benzothiadiazole,5-bromomethyl, 2,1,3-benzothiadiazole, 5-bromomethyl, 5-bromomethyl benzo c 1,2,5-thiadiazole, 5-bromomethyl-2,1,3-benzthiadiazole, 5-bromomethylbenzo-2.1.3-thiadiazole, 5-bromomethylbenzo 2,1,3 thiadiazole
Formula Weight 229.1
Percent Purity 95%
Chemical Name or Material 5-(Bromomethyl)-2, 1, 3-benzothiadiazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.