Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ (±)-5'-Chloro-5'-deoxy-ENBA

Highly selective A1 agonist

$337.50 - $1463.00

Specifications

Purity >98%
Product Type (±)-5'-Chloro-5'-deoxy-ENBA
Molecular Weight (g/mol) 379.84
Inhibitors (±)-5'-Chloro-5'-deoxy-ENBA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
35-761-0R
View Documents
Tocris Bioscience™
3576/10
10 mg
Each for $337.50
missing translation for 'QuantityLeft'
Add to Cart
 
35-765-0R
View Documents
Tocris Bioscience™
3576/50
50 mg
Each for $1,463.00
missing translation for 'QuantityLeft'
Add to Cart
 
Description

Description

Highly selective adenosine A1 receptor agonist (Ki values are 0.51, 1290, 1340 and 2740 nM at A1, A3, A2A and A2B receptors respectively). Reverses formaline-induced nocifensive behavior in mice; antinociceptive.
Specifications

Specifications

>98%
(±)-5'-Chloro-5'-deoxy-ENBA
379.84
(±)-5'-Chloro-5'-deoxy-ENBA
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.