Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Cyanophthalide 98.0+%, TCI America™
SDP

Catalog No. p-7125232 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 82104-74-3
Molecular Formula C9H5NO2
Molecular Weight (g/mol) 159.144
InChI Key XEEGWTLAFIZLSF-UHFFFAOYSA-N
Synonym 5-cyanophthalide, 1-oxo-1,3-dihydroisobenzofuran-5-carbonitrile, 5-cyano phthalide, 5-cyano-3h-isobenzofuranone, 1,3-dihydro-1-oxoisobenzofuran-5-carbonitrile, 1-oxo-1,3-dihydro-2-benzofuran-5-carbonitrile, 1-oxo-3h-isobenzofuran-5-carbonitrile, 1,3-dihydro-1-oxo-5-isobenzofurancarbonitrile, 5-isobenzofurancarbonitrile, 1,3-dihydro-1-oxo, 5-cyanophthalid
PubChem CID 821218
IUPAC Name 1-oxo-3H-2-benzofuran-5-carbonitrile
SMILES C1C2=C(C=CC(=C2)C#N)C(=O)O1

Specifications

Melting Point 204°C
Color White-Yellow
Quantity 5 g
Formula Weight 159.14
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 5-Cyanophthalide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.