Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Fluoro-2-(trifluoromethyl)pyridine, Thermo Scientific™

Catalog No. MAY00193DA
Click to view available options
Quantity:
Packaging:

Chemical Identifiers

CAS

936841-73-5

Molecular Formula

C6H3F4N

Molecular Weight (g/mol)

165.091

InChI Key

URVIJEPCSODFNU-UHFFFAOYSA-N

Synonym

5-fluoro-2-trifluoromethyl pyridine, pubchem23561, 3-fluoro-6-trifluoromethylpyridine, 5-fluoranyl-2-trifluoromethyl pyridine

PubChem CID

26986205

IUPAC Name

5-fluoro-2-(trifluoromethyl)pyridine

SMILES

C1=CC(=NC=C1F)C(F)(F)F

Specifications

Packaging

Amber Glass Bottle

Quantity

1 g

Formula Weight

165.09

Percent Purity

97%

Chemical Name or Material

5-Fluoro-2-(trifluoromethyl)pyridine

Hazard Category

GHS Signal Word: Warning

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.