Learn More
Enzo Life Sciences 5-Iodotubercidin (1mg). CAS: 24386-93-4
SDP
Supplier:  Enzo Life Sciences BMLEI2930001
Encompass_Preferred
Inhibits ERK2 (Ki = 525nM). Also inhibits adenosine kinase (Ki = 30nM), CK1 and CK2 and insulin receptor kinase (IC50 ranging from 0.4-28µM). Alternative name: 4-Amino-5-iodo-7-(beta-D-ribofuranosyl)-pyrrolo 2 3-d pyrimidine. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (25mg/ml) or 100% ethanol (8mg/ml warm). Long Term Storage: -20°C.
Catalog No. 50-201-0674
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.