Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-methyl-1H-benzo[d]imidazole-2-thiol, Thermo Scientific™

Catalog No. SB00799EE
Click to view available options
:
50g

Specifications

CAS 27231-36-3
Assay Percent Range 97%
Molecular Formula C8H8N2S
MDL Number MFCD00010617
Quantity 50g
Synonym 2-mercapto-5-methylbenzimidazole, 5-methyl-1h-benzo d imidazole-2 3h-thione, 5-methyl-1h-benzimidazole-2-thiol, 5-methyl-2-benzimidazolethiol, 5-methyl-2-mercaptobenzimidazole, 5-methyl-1h-1,3-benzodiazole-2-thiol, 2h-benzimidazole-2-thione, 1,3-dihydro-5-methyl, 1,3-dihydro-5-methyl-2h-benzimidazole-2-thione, 5-methyl-1h-benzoimidazole-2-thiol, 5-methylbenzimidazole-2-thiol
InChI Key CWIYBOJLSWJGKV-UHFFFAOYSA-N
SMILES CC1=CC=C2NC(=S)NC2=C1
IUPAC Name 5-methyl-2,3-dihydro-1H-1,3-benzodiazole-2-thione
Molecular Weight (g/mol) 164.23
PubChem CID 712373
Formula Weight 164.23
Percent Purity 97%
Chemical Name or Material 5-Methyl-1H-benzo[d]imidazole-2-thiol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.