Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methyl-2-thiouracil 98.0+%, TCI America™
SDP

Catalog No. p-7132485 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
10 g
25 g

Chemical Identifiers

CAS 636-26-0
Molecular Formula C5H6N2OS
Molecular Weight (g/mol) 142.176
MDL Number MFCD00009777
InChI Key ZLAQATDNGLKIEV-UHFFFAOYSA-N
Synonym 4-Hydroxy-2-mercapto-5-methylpyrimidine, 4-Hydroxy-5-methyl-2-thiopyrimidine
PubChem CID 720471
IUPAC Name 5-methyl-2-sulfanylidene-1H-pyrimidin-4-one
SMILES CC1=CNC(=S)NC1=O

Specifications

Color White
Quantity 10 g
Formula Weight 142.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methyl-2-thiouracil
RTECSNumber XP2108500
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.