Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methylisoxazole-4-carboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. p-7132832 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 42831-50-5
Molecular Formula C5H4NO3
Molecular Weight (g/mol) 126.09
MDL Number MFCD00955655
InChI Key VQBXUKGMJCPBMF-UHFFFAOYSA-M
Synonym 5-methylisoxazole-4-carboxylic acid, 5-methyl-4-isoxazolecarboxylic acid, unii-bl2q213q2m, 4-isoxazolecarboxylic acid, 5-methyl, 5-methylisoxazole-4-carboxylicacid, leflunomide impurity d ep, pubchem8652, leflunomide impurity d, acmc-209jr8, ksc235o3b
PubChem CID 1425240
IUPAC Name 5-methyl-1,2-oxazole-4-carboxylate
SMILES CC1=C(C=NO1)C([O-])=O

Specifications

Melting Point 147°C
Color White-Yellow
Quantity 5 g
Formula Weight 127.10
Percent Purity ≥98.0% (T,HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methylisoxazole-4-carboxylic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.