Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methylpyrazine-2-carboxylic acid, 97%, Thermo Scientific™

Catalog No. SB01107DE
Click to view available options
Quantity:

Chemical Identifiers

CAS

5521-55-1

Molecular Formula

C6H6N2O2

Molecular Weight (g/mol)

138.126

MDL Number

MFCD00068241

InChI Key

RBYJWCRKFLGNDB-UHFFFAOYSA-N

Synonym

5-methyl-2-pyrazinecarboxylic acid, 2-methylpyrazine-5-carboxylic acid, pyrazinecarboxylic acid, 5-methyl, unii-b861rs5nhi, 5-methylpyrazinecarboxylic acid, 5-methyl pyrazine-2-carboxylic acid, 5-methyl-pyrazine-2-carboxylic acid, 2-carboxy-5-methylpyrazine, 2-methyl-5-pyrazinoic acid, b861rs5nhi

PubChem CID

122831

IUPAC Name

5-methylpyrazine-2-carboxylic acid

SMILES

CC1=NC=C(N=C1)C(=O)O

Specifications

CAS Min %

97.0

Assay Percent Range

97%

Quantity

5 g

Formula Weight

138.13

Percent Purity

97%

Chemical Name or Material

5-Methylpyrazine-2-carboxylic acid

Hazard Statement

GHS H Statement:
Causes serious eye damage.

Precautionary Statement

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.