Learn More
TARGETMOL CHEMICALS INC 5-N-ETHYLCARBOXAMIDOADEN 50MG
Supplier: TARGETMOL CHEMICALS INC T7340
Also available in 5 mg 10 mg 25 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 5'-N-Ethylcarboxamidoadenosine (NECA) is an agonist of Adenosine receptor increases cerebral extravasation of fluorescein and low molecular weight dextran independent of blood-brain barrier modulation. purity: 98%
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.