Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Phenoxy-2-furoic acid, ≥97%, Thermo Scientific™

Catalog No. HTS06019DA
Click to view available options
Quantity:

Chemical Identifiers

CAS

60698-32-0

Molecular Formula

C11H8O4

Molecular Weight (g/mol)

204.18

MDL Number

MFCD01925067

InChI Key

QQKXTFLRAZKJJO-UHFFFAOYSA-N

Synonym

5-phenoxy-2-furoic acid, 2-furancarboxylic acid,5-phenoxy, 5-phenoxy-furan-2-carboxylic acid, maybridge3_002285, 5-phenoxypyromucic acid

PubChem CID

2812790

IUPAC Name

5-phenoxyfuran-2-carboxylic acid

SMILES

OC(=O)C1=CC=C(OC2=CC=CC=C2)O1

Specifications

CAS Min %

97.0

Assay Percent Range

97%

Packaging

Amber glass bottle

Quantity

1 g

Formula Weight

204.18

Percent Purity

97%

Chemical Name or Material

5-Phenoxy-2-furoic acid

Hazard Category

GHS Signal Word: Danger

Hazard Statement

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

Precautionary Statement

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.