Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ 50 g Aluminum Weight Hanger
SDPSCI_ED

Easily store and use your slotted weights.

Supplier:  United Scientific™ WHNG50A

Catalog No. S139465


missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
This item is not returnable. View return policy

Description

Description

  • Aluminum weight hanger for use with slotted weights
  • Stem length is 14 cm
  • Weight is 50 g
Specifications

Specifications

Weight Hanger
Aluminum
50 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

Warning: Cancer and reproductive harm

Prop 65