Learn More
Chemglass Life Sciences 50000mL Glas-Col Heating Mantle, Series TM, Spherical, 3 @ 1000 Watts
SDP
Supplier:  Chemglass Life Sciences CG1200014
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
50000mL Glas-Col Heating Mantle, Series TM, Spherical, 3 @ 1000 Watts. The Glas-Col Series TM spherical flask mantle covers only the flasks bottom half, letting you see its full contents. Larger sizes have multiple circuits for ease of temperature control when a flask is less than half full. The Series TM mantle is available with a bottom hole for flask drain on a special order basis. Terminal box models are available with switches and pilot lights. The 50 Liter size is furnished with a terminal box instead of a cord.
Catalog No. CG1200014
missing translation for 'QuantityLeft'