Learn More
Chemglass Life Sciences 500mL Peptide Synthesis Vessel, Solid Phase, Medium Frit, GL 32 Thread
SDP
Supplier:  Chemglass Life Sciences CG186106
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
500mL Peptide Synthesis Vessel, Solid Phase, Medium Frit, GL 32 Thread. Solid phase peptide synthesis vessel having a medium or coarse porosity fritted glass resin support. Top of the vessel has a GL thread and is supplied complete with a PTFE lined PBT screw cap. Side port and lower PTFE stopcock have a 2mm bore with the arms being 8mm I.D. Vessels are available in sizes other than those listed. Please call for details.
Catalog No. CG186106