Learn More
Ace Glass, Inc. 500mL jacketed reactor body only, no bottom valve stem or#7 nylon bushings
Supplier:  Ace Glass, Inc. 654140
Encompass_Preferred
500mL jacketed reactor body with three angled 24/40 joints and two #7 Ace-Threds. Valve stem and #7 bushings are not included.
Catalog No. 50-978-818
missing translation for 'QuantityLeft'
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.