Learn More
Chemglass Life Sciences 50L Process Reactor, Complete, Electric Motor, 90.8" OAH with Motor, 18.1" Clearance Below Valve
SDP
Supplier:  Chemglass Life Sciences CG1972650
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
50L Process Reactor, Complete, Electric Motor, 90.8" OAH with Motor, 18.1" Clearance Below Valve.Features:• Motor Mount Arm Can Unlock And Swing Out Giving Greater Access To Reactor• Smaller Footprint Support Frame With Oversized Rugged S.S. Uprights And Heavy Duty PTFE Coated Aluminum Fittings• Electric And XP Motor Has Separate Controller With Digital Display• Reactor Support Plate Holds Reactor Securely And Allows Easy Installation/Removal Of Reactor By One Person• Lower Safety Ring And Lower Mantle Base• Heavy Duty Lockable Casters• Electric Or Air Motor Optional XP Motor Available• Open Frame Design Provides Excellent Accessibility• 7-Neck Lid With Large Powder Addition Port• Ridge Clamp Accepts 1/2” Rod And Adds Additional Support For Large Glassware• #304 S.S. Cooling Coil PTFE Coated • Digital Temperature Controller• Zero Dead Space Detachable Drain Valve Has Knob With Laser Etched Directional Arrows For Open And Close
Catalog No. CG1972650
missing translation for 'QuantityLeft'