Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ 5J 4
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
5J 4 is a calcium release-activated calcium (CRAC) channel blocker. Blocks store-operated calcium entry (SOCE) in HeLa-O+S cells and Th17 cells in vitro. Ameliorates symptoms and reduces CNS infiltration by mononuclear cells in a mouse model of experimental autoimmune encephalomyelitis (EAE) in vivo.
Specifications
Specifications
Chemical Name or Material | N-[[(6-Hydroxy-1-naphthalenyl)amino]thioxomethyl]-2-furancarboxamide |
CAS | 827001-82-1 |
Quantity | 10 mg |
Target | Other Calcium Channel Blockers |
Molecular Formula | C16H12N2O3S |
Formula Weight | 312.34 |
Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction