Learn More
Sklar Instruments Scissors, Sklar Tungsten Carbide Mayo-Stille Dissecting Scissors, Sklar 6-3/4 in., Blunt/Blunt, Straight, Smooth, Tungsten Carbide / Stainless Steel. EA.
SDP
Supplier:  Sklar Instruments 163557
Encompass_Preferred
Scissors, Sklar Edge Tungsten Carbide Mayo-Stille Dissecting Scissors, Sklar, 6-3/4 in., Blunt/Blunt, Straight, Smooth, Tungsten Carbide / Stainless Steel. EA. Sklar Edge Tungsten Carbide Mayo-Stille Dissecting Scissors feature tungsten carbide inserts that are permanently bonded to the scissor blade. This provides an exceptionally hard and durable cutting edge and increases the overall life the instrument. Mayo-Stille Dissecting Scissors have blunt tips and are available curved or straight. The edges are rounded which permits deeper dissection of tough tissue. Sklar Edge scissors are differentiated from other scissors by the handle of two gold coated finger rings.
Catalog No. 50-109-3570
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.