Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-(4-Aminophenyl)-4,5-dihydro-5-methyl-3(2H)-pyridazinone 98.0+%, TCI America™
SDP

Catalog No. p-7118145 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 36725-28-7
Molecular Formula C11H13N3O
Molecular Weight (g/mol) 203.245
MDL Number MFCD01692712
InChI Key GDMRFHZLKNYRRO-UHFFFAOYSA-N
PubChem CID 161928
IUPAC Name 3-(4-aminophenyl)-4-methyl-4,5-dihydro-1H-pyridazin-6-one
SMILES CC1CC(=O)NN=C1C2=CC=C(C=C2)N

Specifications

Melting Point 196°C
Quantity 1 g
Formula Weight 203.25
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 6-(4-Aminophenyl)-4,5-dihydro-5-methyl-3(2H)-pyridazinone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.