Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6,7-Dimethoxy-3,4-dihydroisoquinoline 98.0+%, TCI America™
SDP

Catalog No. p-7128629 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 3382-18-1
Molecular Formula C11H13NO2
Molecular Weight (g/mol) 191.23
MDL Number MFCD00266748
InChI Key NSLJVQUDZCZJLK-UHFFFAOYSA-N
Synonym 6,7-dimethoxy-3,4-dihydro-isoquinoline, 3,4-dihydro-6,7-dimethoxyisoquinoline, isoquinoline, 3,4-dihydro-6,7-dimethoxy, pubchem12828, ksc571e8h, 3,4-dihydro-6,7-dimethoxy-isoquinoline, isoquinoline,3,4-dihydro-6,7-dimethoxy, isoquinoline, 3,4-dihydro-6,7-dimethoxy-, hydrochloride 1:1
PubChem CID 30058
IUPAC Name 6,7-dimethoxy-3,4-dihydroisoquinoline
SMILES COC1=CC2=C(C=NCC2)C=C1OC

Specifications

Melting Point 49°C
Boiling Point 160°C
Quantity 1 g
Formula Weight 191.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 6,7-Dimethoxy-3,4-dihydroisoquinoline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.