Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Aminofluorescein (isomer II) 95.0+%, TCI America™
SDP

Catalog No. p-7117096 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 51649-83-3
Molecular Formula C20H13NO5
Molecular Weight (g/mol) 347.326
MDL Number MFCD00005051
InChI Key YOAWSYSKQHLFPM-UHFFFAOYSA-N
Synonym 6-aminofluorescein, fluoresceinamine isomer ii, 2-aminofluorescein, 6-amino-3',6'-dihydroxy-3h-spiro isobenzofuran-1,9'-xanthen-3-one, 6-amino-3',6'-dihydroxy-3h-spiro 2-benzofuran-1,9'-xanthen-3-one, 5-amino-3',6'-dihydroxyspiro 2-benzofuran-3,9'-xanthene-1-one, 6-amino-3',6'-dihydroxyspiro 2-benzofuran-1,9'-xanthene-3-one, 6-amino-3',6'-dihydroxyspiro isobenzofuran-1 3h ,9'-9h xanthene-3-one, spiro isobenzofuran-1 3h ,9'-9h xanthen-3-one, 6-amino-3',6'-dihydroxy
PubChem CID 103924
IUPAC Name 5-amino-3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one
SMILES C1=CC2=C(C=C1N)C3(C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O)OC2=O

Specifications

Melting Point 285°C
Quantity 1 g
Formula Weight 347.33
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Aminofluorescein (isomer II)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.