Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Aminonicotinamide, MP Biomedicals™

Catalog No. ICN15010305 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:

Chemical Identifiers

CAS

329-89-5

Molecular Formula

C6H7N3O

Molecular Weight (g/mol)

137.14

MDL Number

MFCD00006327

InChI Key

ZLWYEPMDOUQDBW-UHFFFAOYSA-N

Synonym

6-aminonicotinamide, 2-amino-5-carbamoylpyridine, 3-pyridinecarboxamide, 6-amino, 6-amino-3-pyridinecarboxamide, 6-aminonicotinic acid amide, 6-aminonikotinsaeureamid, nicotinamide, 6-amino, aminonicotinamide, 6-an, 6-amino-nicotinsaeureamid

PubChem CID

9500

ChEBI

CHEBI:74514

IUPAC Name

6-aminopyridine-3-carboxamide

SMILES

NC(=O)C1=CC=C(N)N=C1

Specifications

Quantity

5 g

Formula Weight

137.1

Physical Form

Solid

Chemical Name or Material

6-Aminonicotinamide

Recommended Storage

Store at 2° to 8°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.