Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Bromoindazole 98.0+%, TCI America™
SDP

Catalog No. p-7123335 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g
25 g

Chemical Identifiers

CAS 79762-54-2
Molecular Formula C7H5BrN2
Molecular Weight (g/mol) 197.04
MDL Number MFCD03265457
InChI Key WMKDUJVLNZANRN-UHFFFAOYSA-N
Synonym 6-bromoindazole, 6-bromo-2h-indazole, 1h-indazole, 6-bromo, 1h-indazole,6-bromo, pubchem7831, zlchem 1237, 6-bromo-1-h-indazole, 6-bromanyl-1h-indazole, acmc-209pi5
PubChem CID 17842471
IUPAC Name 6-bromo-1H-indazole
SMILES BrC1=CC=C2C=NNC2=C1

Specifications

Melting Point 183°C
Quantity 1 g
Formula Weight 197.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromoindazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.