Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Bromoquinoxaline, 97%, Thermo Scientific™

Catalog No. p-2703317
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 50998-17-9
Molecular Formula C8H5BrN2
Molecular Weight (g/mol) 209.05
MDL Number MFCD00837757
InChI Key NOYFLUFQGFNMRB-UHFFFAOYSA-N
Synonym 6-bromo-quinoxaline, quinoxaline, 6-bromo, 6-bromobenzopyrazine, acmc-209krf, 6-bromoquinoxaline, ksc269e9f
PubChem CID 610939
IUPAC Name 6-bromoquinoxaline
SMILES BrC1=CC=C2N=CC=NC2=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 209.05
Percent Purity 97%
Chemical Name or Material 6-Bromoquinoxaline
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.