Learn More
TARGETMOL CHEMICALS INC 6-CHLOROMELATONIN 5MG
Supplier:  TARGETMOL CHEMICALS INC T22530
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. 6-Chloromelatonin is a potent melatonin receptor agonist a 5-methoxyindole compound that competes for the same presynaptic melatonin receptor sites in the rabbit retina and inhibits the calcium-dependent release of [3H]dopamine with a higher metabolic stability than melatonin. Chloromelatonin competes for [3H]melatonin binding to MT2 receptors (pKi=9.77) and may be used to study insomnia and sleep disorders associated with depression. purity: 99%
Catalog No. 50-258-0947
missing translation for 'QuantityLeft'