Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Methoxy-8-nitroquinoline 98.0+%, TCI America™
SDP

Catalog No. p-7132993 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 85-81-4
Molecular Formula C10H8N2O3
Molecular Weight (g/mol) 204.185
MDL Number MFCD00006802
InChI Key MIMUSZHMZBJBPO-UHFFFAOYSA-N
Synonym quinoline, 6-methoxy-8-nitro, 6-methoxy-8-nitro-quinoline, 8-nitro-6-methoxyquinoline, methyl 8-nitro-6-quinolyl ether, unii-ats412o1xo, ats412o1xo, pubchem9433, acmc-209q7l, 5-21-03-00248 beilstein handbook reference, ksc655o7n
PubChem CID 6822
IUPAC Name 6-methoxy-8-nitroquinoline
SMILES COC1=CC(=C2C(=C1)C=CC=N2)[N+](=O)[O-]

Specifications

Melting Point 161°C
Color Yellow
Quantity 5 g
Formula Weight 204.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Methoxy-8-nitroquinoline
RTECSNumber VC0365000
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.