Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Methoxyflavanone 98.0+%, TCI America™
SDP

Catalog No. p-7132710 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 3034-04-6
Molecular Formula C16H14O3
Molecular Weight (g/mol) 254.29
MDL Number MFCD00017484
InChI Key YURQMHCZHLMHIB-UHFFFAOYNA-N
Synonym 6-methoxyflavanone, 6-methoxy-2-phenylchroman-4-one, 6-methoxy-2-phenyl-chroman-4-one, 6-methoxy-2-phenyl-2,3-dihydro-1-benzopyran-4-one, flavanone, 6-methoxy, acmc-1adv8, 6-methoxy-2-phenyl-2,3-dihydro-4h-chromen-4-one, 6-methoxy-2-phenyl-2,3-dihydro-4h-chromen-4-one #, 2,3-dihydro-6-methoxy-2-phenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 2,3-dihydro-6-methoxy-2-phenyl
PubChem CID 97860
IUPAC Name 6-methoxy-2-phenyl-2,3-dihydrochromen-4-one
SMILES COC1=CC2=C(C=C1)OC(CC2=O)C3=CC=CC=C3

Specifications

Melting Point 142°C
Color White-Yellow
Quantity 1 g
Formula Weight 254.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Methoxyflavanone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.